Assay Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029432.1 | NP_083708.1 | 33 - 34 | 3121 | 63 | |
| GenBank mRNA | AK133294.1 | - | 33 - 34 | 3263 | 63 | |
| AK143611.1 | - | 33 - 34 | 3146 | 63 | ||
| BC052447.1 | - | 33 - 34 | 3121 | 6408647 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930402H24Rik
Genomic Map