Assay Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029432.1 | NP_083708.1 | 7 - 8 | 758 | 58 | |
| GenBank mRNA | AK015050.1 | - | 7 - 8 | 760 | 58 | |
| AK089462.1 | - | 7 - 8 | 782 | 58 | ||
| AK133294.1 | - | 7 - 8 | 900 | 58 | ||
| AK143611.1 | - | 7 - 8 | 783 | 58 | ||
| AK158367.1 | - | 7 - 8 | 780 | 58 | ||
| BC052447.1 | - | 7 - 8 | 758 | 6408647 | 58 | |
More Information
Set Membership: |
|

4930402H24Rik
Genomic Map