Assay Details
| Gene Symbol: Myo1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053214.2 | NP_444444.2 | 15 - 16 | 1755 | 110 | |
| GenBank mRNA | AK036840.1 | - | 5 - 6 | 1588 | 110 | |
| AK131188.1 | - | 5 - 6 | 448 | 110 | ||
| AK153908.1 | - | 15 - 16 | 1718 | 110 | ||
| AK158333.1 | - | 15 - 16 | 1670 | 110 | ||
| BC046502.1 | - | 15 - 16 | 1743 | 4167558 | 110 | |
More Information
Set Membership: |
|

Myo1f
Genomic Map