Assay Details
| Gene Symbol: H2-Ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207105.3 | NP_996988.2 | 1 - 2 | 177 | 60 | |
| GenBank mRNA | AK077436.1 | - | 1 - 2 | 165 | 60 | |
| AK170370.1 | - | 1 - 2 | 159 | 60 | ||
| BY205244.1 | - | 1 - 2 | 196 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

H2-Ab1
Genomic Map