Assay Details
| Gene Symbol: H2-Ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207105.3 | NP_996988.2 | 4 - 5 | 844 | 139 | |
| GenBank mRNA | AK077436.1 | - | 4 - 5 | 832 | 139 | |
| AK170370.1 | - | 4 - 5 | 826 | 139 | ||
| CK330363.1 | - | 2 - 3 | 342 | 139 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

H2-Ab1
Genomic Map