Assay Details
| Gene Symbol: Tmem62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175285.3 | NP_780494.1 | 1 - 2 | 417 | 70 | |
| GenBank mRNA | AK046793.1 | - | 1 - 2 | 216 | 70 | |
| BC070410.1 | - | 1 - 2 | 417 | 5708443 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem62
Genomic Map