Assay Details
| Gene Symbol: Itgax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021334.2 | NP_067309.1 | 18 - 19 | 2339 | 64 | |
| GenBank mRNA | AF211864.1 | - | 18 - 19 | 2339 | 64 | |
| AK131133.1 | - | 18 - 19 | 2345 | 64 | ||
| AK155816.1 | - | 18 - 19 | 2361 | 64 | ||
| AK170432.1 | - | 18 - 19 | 2357 | 64 | ||
| BC042840.1 | - | 18 - 19 | 2338 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgax
Genomic Map