Assay Details
| Gene Symbol: Sestd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175465.6 | NP_780674.1 | 13 - 14 | 1497 | 64 | |
| GenBank mRNA | BC011970.1 | - | 3 - 4 | 577 | 64 | |
| BC013245.1 | - | 14 - 15 | 1578 | 64 | ||
| BC044853.1 | - | 13 - 14 | 1497 | 3978465 | 64 | |
| BC048358.1 | - | 1 - 2 | 50 | 64 | ||
| CF175139.1 | - | 3 - 4 | 347 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sestd1
Genomic Map