Assay Details
| Gene Symbol: Sestd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175465.6 | NP_780674.1 | 15 - 16 | 1700 | 97 | |
| GenBank mRNA | BC011970.1 | - | 5 - 6 | 780 | 97 | |
| BC038005.1 | - | 2 - 3 | 211 | 97 | ||
| BC044853.1 | - | 15 - 16 | 1700 | 3978465 | 97 | |
| BC048358.1 | - | 3 - 4 | 253 | 97 | ||
| CF175139.1 | - | 5 - 6 | 550 | 97 | ||
| CN664477.1 | - | 1 - 2 | 105 | 97 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sestd1
Genomic Map