Assay Details
| Gene Symbol: Sestd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175465.6 | NP_780674.1 | 2 - 3 | 109 | 89 | |
| GenBank mRNA | AK047939.1 | - | 2 - 3 | 107 | 89 | |
| BB647911.1 | - | 2 - 3 | 93 | 89 | ||
| BC013245.1 | - | 3 - 4 | 190 | 89 | ||
| BC044853.1 | - | 2 - 3 | 109 | 3978465 | 89 | |
| BY106142.1 | - | 2 - 3 | 142 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sestd1
Genomic Map