Assay Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017461.2 | NP_059489.2 | 10 - 11 | 1054 | 71 | |
| GenBank mRNA | AV507599.1 | - | 2 - 3 | 187 | 71 | |
| AY974598.1 | - | 2 - 3 | 174 | 71 | ||
| BC140998.1 | - | 10 - 11 | 1055 | 9055610 | 71 | |
| BC145081.1 | - | 10 - 11 | 1055 | 9053629 | 71 | |
| BX523183.1 | - | 3 - 4 | 370 | 71 | ||
| M37030.1 | - | 10 - 11 | 1058 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178127 | M37030.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178127 | NM_017461.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178127 | AW487826.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963764 | BX523183.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963764 | M37030.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963764 | NM_017461.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963764 | AV507599.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963764 | BC140998.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | AF250729.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | AK012392.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | AK017691.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | AK051683.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | AK132316.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | AK133622.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | AK136921.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | BC016244.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663732 | NM_029756.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764842 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169060 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178127 | M37030.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178127 | NM_017461.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178127 | AW487826.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963764 | BX523183.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963764 | M37030.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963764 | NM_017461.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963764 | AV507599.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963764 | BC140998.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504623 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169060 | - | - |
| Whole Mouse Genome G4122A | A_52_P91527 | M37030.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P91527 | NM_017461.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P91527 | BC140998.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P91527 | BC145081.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124935 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169060 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06040798 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06283358 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06283362 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06040793 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06602738 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06040802 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06283360 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06040794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06331572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06056530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06056519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06040796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06040802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06207469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06040804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06198306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06342877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06283360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06503791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06602739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06283362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504628 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568214 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568210 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568208 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449898_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568202 | - | - |
| Mouse Genome 430 2.0 Array | 1449898_at | - | - |
| Mouse Genome 430 2.0 Array | 1449898_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54204_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54204_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700036253.mm.1 | NM_017461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036253.mm.1 | NM_017461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036253.mm.1 | TC0700004318.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036255.mm.1 | NM_017461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036255.mm.1 | NM_017461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036255.mm.1 | TC0700004318.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036257.mm.1 | NM_017461.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036257.mm.1 | NM_017461.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036257.mm.1 | TC0700004318.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449898_PM_at | - | - |
| Mouse Expression Set 430 | 1449898_at | - | - |
| Murine Genome U74v2 Set | 102962_PM_at | - | - |
| Murine Genome U74v2 Set | 102962_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449898_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449898_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449898_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449898_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 864974 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3290546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225633 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225633 | - | - |
| mouse-6 v1.1 (Illumina) | scl054204.2_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl054204.2_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225633 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225633 | - | - |

Sept1
Genomic Map