Assay Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017461.2 | NP_059489.2 | 4 - 5 | 345 | 82 | |
| GenBank mRNA | BB610735.1 | - | 4 - 5 | 345 | 82 | |
| BC140998.1 | - | 4 - 5 | 346 | 9055610 | 82 | |
| BC145081.1 | - | 4 - 5 | 346 | 9053629 | 82 | |
| M37030.1 | - | 4 - 5 | 352 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sept1
Genomic Map