Assay Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017461.2 | NP_059489.2 | 5 - 6 | 477 | 69 | |
| GenBank mRNA | BB610735.1 | - | 5 - 6 | 477 | 69 | |
| BC140998.1 | - | 5 - 6 | 478 | 9055610 | 69 | |
| BC145081.1 | - | 5 - 6 | 478 | 9053629 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept1
Genomic Map