Assay Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017461.2 | NP_059489.2 | 8 - 9 | 786 | 84 | |
| GenBank mRNA | BC140998.1 | - | 8 - 9 | 787 | 9055610 | 84 |
| BC145081.1 | - | 8 - 9 | 787 | 9053629 | 84 | |
| BX523183.1 | - | 1 - 2 | 102 | 84 | ||
| M37030.1 | - | 8 - 9 | 790 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sept1
Genomic Map