Assay Details
| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172748.2 | NP_766336.2 | 7 - 8 | 1685 | 66 | |
| GenBank mRNA | AK028405.1 | - | 7 - 8 | 66 | ||
| AK031431.1 | - | 5 - 6 | 1078 | 66 | ||
| AK162161.1 | - | 1 - 2 | 64 | 66 | ||
| BC053109.1 | - | 3 - 4 | 678 | 66 | ||
| BC059812.1 | - | 7 - 8 | 1685 | 6401846 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK162161.1 transcripts.
Set Membership: |
|

Fbxl19
Genomic Map