Assay Details
| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081087.1 | NP_001074556.1 | 2 - 3 | 1349 | 138 | |
| GenBank mRNA | AK132622.1 | - | 2 - 3 | 362 | 138 | |
| BC150698.1 | - | 2 - 3 | 1434 | 9087609 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl41
Genomic Map