Assay Details
| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081087.1 | NP_001074556.1 | 3 - 4 | 1449 | 165 | |
| GenBank mRNA | AK132622.1 | - | 3 - 4 | 462 | 165 | |
| BC150698.1 | - | 3 - 4 | 1534 | 9087609 | 165 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl41
Genomic Map