Assay Details
| Gene Symbol: 5830411J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017919.1 | - | 1 | 1566 | 133 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tousled-like kinase 1 | Tlk1 | 4930545J15Rik | Sense | Chr. 2 - 70712407 - 70825480 on NCBI Build GRCm38 | |
| Downstream | methyltransferase like 8 | Mettl8 |
BC004636, Tip |
141.1kb | Sense | Chr. 2 - 70964558 - 71055643 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830411J07Rik
Genomic Map