Assay Details
| Gene Symbol: Ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110145.1 | NP_001103615.1 | 2 - 3 | 233 | 101 | |
| NM_009278.4 | NP_033304.1 | 2 - 3 | 260 | 101 | ||
| GenBank mRNA | AK017822.1 | - | 2 - 3 | 174 | 101 | |
| AK032940.1 | - | 2 - 3 | 177 | 101 | ||
| AK088650.1 | - | 2 - 3 | 216 | 101 | ||
| AK088677.1 | - | 2 - 3 | 167 | 101 | ||
| AK134864.1 | - | 2 - 3 | 198 | 101 | ||
| AK141487.1 | - | 2 - 3 | 155 | 101 | ||
| AK168828.1 | - | 2 - 3 | 183 | 101 | ||
| BC003820.1 | - | 2 - 3 | 120 | 3593742 | 101 | |
| BC106150.1 | - | 2 - 3 | 113 | 3483153 | 101 | |
| L00993.1 | - | 2 - 3 | 196 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017822.1, AK032940.1, AK088650.1, AK088677.1, AK141487.1, AK168828.1, BC003820.1, BC106150.1, L00993.1, NM_001110145.1, NM_009278.4 transcripts.
Set Membership: |
|

Ssb
Genomic Map