Assay Details
| Gene Symbol: Ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110145.1 | NP_001103615.1 | 9 - 10 | 982 | 129 | |
| NM_009278.4 | NP_033304.1 | 9 - 10 | 1009 | 129 | ||
| GenBank mRNA | AK032940.1 | - | 9 - 10 | 926 | 129 | |
| AK088650.1 | - | 9 - 10 | 965 | 129 | ||
| AK088677.1 | - | 9 - 10 | 129 | |||
| AK134864.1 | - | 9 - 10 | 947 | 129 | ||
| AK141487.1 | - | 9 - 10 | 904 | 129 | ||
| AK168828.1 | - | 9 - 10 | 932 | 129 | ||
| BC003820.1 | - | 9 - 10 | 869 | 3593742 | 129 | |
| BC106150.1 | - | 9 - 10 | 862 | 3483153 | 129 | |
| L00993.1 | - | 9 - 10 | 945 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssb
Genomic Map