Assay Details
| Gene Symbol: Scn7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009135.2 | NP_033161.2 | 18 - 19 | 3239 | 83 | |
| GenBank mRNA | BC145273.1 | - | 18 - 19 | 3095 | 83 | |
| L36179.1 | - | 18 - 19 | 3275 | 83 | ||
More Information
Set Membership: |
|

Scn7a
Genomic Map