Assay Details
| Gene Symbol: Scn7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009135.2 | NP_033161.2 | 7 - 8 | 1089 | 73 | |
| GenBank mRNA | AK142964.1 | - | 7 - 8 | 1094 | 73 | |
| BC145273.1 | - | 7 - 8 | 946 | 73 | ||
| L36179.1 | - | 7 - 8 | 1125 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scn7a
Genomic Map