Assay Details
| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141931.1 | NP_001135403.1 | 10 - 11 | 1187 | 82 | |
| NM_001141932.1 | NP_001135404.1 | 11 - 12 | 1235 | 82 | ||
| NM_020296.2 | NP_064692.2 | 10 - 11 | 1380 | 82 | ||
| GenBank mRNA | AB026569.1 | - | 10 - 11 | 1103 | 82 | |
| AK133054.1 | - | 10 - 11 | 908 | 82 | ||
| AK135280.1 | - | 10 - 11 | 1067 | 82 | ||
| AK145897.1 | - | 10 - 11 | 1189 | 82 | ||
| AK147109.1 | - | 10 - 11 | 1381 | 82 | ||
| AK161102.1 | - | 11 - 12 | 1185 | 82 | ||
| AK161416.1 | - | 6 - 7 | 517 | 82 | ||
| AK178547.1 | - | 5 - 6 | 267 | 82 | ||
| AK210454.1 | - | 5 - 6 | 296 | 82 | ||
| AK218973.1 | - | 2 - 3 | 126 | 82 | ||
| BC016501.1 | - | 10 - 11 | 1368 | 4527742 | 82 | |
| BC056609.1 | - | 8 - 9 | 627 | 82 | ||
| BC057866.1 | - | 11 - 12 | 1267 | 5327143 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbms1
Genomic Map