Assay Details
| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141931.1 | NP_001135403.1 | 11 - 12 | 1300 | 104 | |
| NM_001141932.1 | NP_001135404.1 | 12 - 13 | 1348 | 104 | ||
| NM_020296.2 | NP_064692.2 | 11 - 12 | 1493 | 104 | ||
| GenBank mRNA | AB026569.1 | - | 11 - 12 | 1216 | 104 | |
| AK133054.1 | - | 11 - 12 | 1021 | 104 | ||
| AK135280.1 | - | 11 - 12 | 1180 | 104 | ||
| AK145897.1 | - | 11 - 12 | 1302 | 104 | ||
| AK147109.1 | - | 11 - 12 | 1494 | 104 | ||
| AK161102.1 | - | 12 - 13 | 1298 | 104 | ||
| AK161416.1 | - | 7 - 8 | 630 | 104 | ||
| BC016501.1 | - | 11 - 12 | 1481 | 4527742 | 104 | |
| BC056609.1 | - | 9 - 10 | 740 | 104 | ||
| BC057866.1 | - | 12 - 13 | 1380 | 5327143 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbms1
Genomic Map