Assay Details
| Gene Symbol: Pde3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011055.2 | NP_035185.2 | 1 - 2 | 1211 | 74 | |
| GenBank mRNA | AF547435.1 | - | 1 - 2 | 1184 | 74 | |
| AJ132271.1 | - | 1 - 2 | 914 | 74 | ||
| BY745829.1 | - | 1 - 2 | 114 | 74 | ||
| X95521.1 | - | 1 - 2 | 56 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde3b
Genomic Map