Assay Details
Gene Symbol: Ddrgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029832.2 | NP_084108.1 | 2 - 3 | 376 | 80 | |
GenBank mRNA | AK011170.1 | - | 2 - 3 | 307 | 80 | |
BC115616.2 | - | 2 - 3 | 376 | 40102754 | 80 | |
BC115617.1 | - | 2 - 3 | 376 | 40102757 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
