Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200015998.mm.1 |
NM_001206382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015998.mm.1 |
NM_001206382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015998.mm.1 |
NM_001206385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015998.mm.1 |
NM_001206385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015998.mm.1 |
NM_144888.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015998.mm.1 |
NM_144888.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015998.mm.1 |
TC0200002012.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015995.mm.1 |
NM_144888.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015995.mm.1 |
NM_144888.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015995.mm.1 |
TC0200002012.mm.1 |
MTA TC |