Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200010620.mm.1 |
NM_001289678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010620.mm.1 |
NM_028733.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010620.mm.1 |
NM_030880.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010620.mm.1 |
TC0200001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010611.mm.1 |
NM_001289678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010611.mm.1 |
NM_030880.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010611.mm.1 |
TC0200001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010613.mm.1 |
NM_001289678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010613.mm.1 |
NM_030880.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010613.mm.1 |
TC0200001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010612.mm.1 |
NM_001289678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010612.mm.1 |
NM_030880.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010612.mm.1 |
TC0200001374.mm.1 |
MTA TC |