Assay Details
| Gene Symbol: Ckmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009897.2 | NP_034027.1 | 7 - 8 | 1217 | 74 | |
| GenBank mRNA | AK005076.1 | - | 7 - 8 | 1216 | 74 | |
| AK131953.1 | - | 7 - 8 | 1221 | 74 | ||
| BC025976.1 | - | 8 - 9 | 1132 | 4216758 | 74 | |
| Z13968.1 | - | 7 - 8 | 1217 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ckmt1
Genomic Map