Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 13 - 14 | 2919 | 92 | |
| NM_013735.4 | NP_038763.3 | 13 - 14 | 2919 | 92 | ||
| GenBank mRNA | AJ414734.1 | - | 12 - 13 | 2780 | 92 | |
| BC079906.1 | - | 13 - 14 | 2853 | 30639375 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map