Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 15 - 16 | 3177 | 82 | |
| NM_013735.4 | NP_038763.3 | 15 - 16 | 3177 | 82 | ||
| GenBank mRNA | AJ414734.1 | - | 14 - 15 | 3038 | 82 | |
| AK045851.1 | - | 14 - 15 | 2972 | 82 | ||
| BC035206.1 | - | 3 - 4 | 407 | 3979746 | 82 | |
| BC079906.1 | - | 15 - 16 | 3111 | 30639375 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map