Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 18 - 19 | 3928 | 66 | |
| NM_013735.4 | NP_038763.3 | 18 - 19 | 3928 | 66 | ||
| GenBank mRNA | AJ414734.1 | - | 17 - 18 | 3771 | 66 | |
| AK029832.1 | - | 325 | 66 | |||
| BC035206.1 | - | 6 - 7 | 1140 | 3979746 | 66 | |
| BC079906.1 | - | 18 - 19 | 3862 | 30639375 | 66 | |
| CA315900.1 | - | 2 - 3 | 599 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map