Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 1 - 2 | 139 | 83 | |
| NM_013735.4 | NP_038763.3 | 1 - 2 | 139 | 83 | ||
| GenBank mRNA | AK045077.1 | - | 1 - 2 | 94 | 83 | |
| AK148123.1 | - | 1 - 2 | 102 | 82 | ||
| BC037378.1 | - | 1 - 2 | 101 | 83 | ||
| BC079906.1 | - | 1 - 2 | 73 | 30639375 | 83 | |
| BY777325.1 | - | 1 - 2 | 140 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK148123.1, BC079906.1, NM_001290830.1, NM_013735.4 transcripts.
Set Membership: |
|

Trp53bp1
Genomic Map