Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013735.4 | NP_038763.3 | 20 - 21 | 4351 | 59 | |
| GenBank mRNA | AJ414734.1 | - | 19 - 20 | 4194 | 59 | |
| AK029832.1 | - | 748 | 59 | |||
| AK036324.1 | - | 1 - 2 | 156 | 59 | ||
| AK132323.1 | - | 2 - 3 | 384 | 59 | ||
| BC035206.1 | - | 8 - 9 | 1563 | 3979746 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map