Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 21 - 22 | 4832 | 59 | |
| NM_013735.4 | NP_038763.3 | 22 - 23 | 4982 | 59 | ||
| GenBank mRNA | AJ414734.1 | - | 21 - 22 | 4825 | 59 | |
| AK029832.1 | - | 1379 | 59 | |||
| AK036324.1 | - | 3 - 4 | 787 | 59 | ||
| AK132323.1 | - | 4 - 5 | 1015 | 59 | ||
| BC035206.1 | - | 10 - 11 | 2194 | 3979746 | 59 | |
| BC079906.1 | - | 21 - 22 | 4766 | 30639375 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map