Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 2 - 3 | 318 | 68 | |
| NM_013735.4 | NP_038763.3 | 2 - 3 | 318 | 68 | ||
| GenBank mRNA | AJ414734.1 | - | 1 - 2 | 182 | 68 | |
| AK045851.1 | - | 2 - 3 | 233 | 68 | ||
| AK148123.1 | - | 2 - 3 | 281 | 68 | ||
| BC037378.1 | - | 2 - 3 | 280 | 68 | ||
| BC079906.1 | - | 2 - 3 | 252 | 30639375 | 68 | |
| BY777325.1 | - | 2 - 3 | 319 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map