Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 7 - 8 | 915 | 94 | |
| NM_013735.4 | NP_038763.3 | 7 - 8 | 915 | 94 | ||
| GenBank mRNA | AJ414734.1 | - | 6 - 7 | 776 | 94 | |
| AK045851.1 | - | 7 - 8 | 830 | 94 | ||
| AK148123.1 | - | 7 - 8 | 875 | 94 | ||
| BC079906.1 | - | 7 - 8 | 849 | 30639375 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map