Assay Details
| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 9 - 10 | 1205 | 101 | |
| NM_013735.4 | NP_038763.3 | 9 - 10 | 1205 | 101 | ||
| GenBank mRNA | AJ414734.1 | - | 8 - 9 | 1066 | 101 | |
| AK045851.1 | - | 9 - 10 | 1120 | 101 | ||
| AK148123.1 | - | 9 - 10 | 1165 | 101 | ||
| BC079906.1 | - | 9 - 10 | 1139 | 30639375 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp1
Genomic Map