Assay Details
| Gene Symbol: Supt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178652.2 | NP_848767.1 | 9 - 10 | 1059 | 86 | |
| GenBank mRNA | AK050321.1 | - | 9 - 10 | 906 | 86 | |
| AK075916.1 | - | 9 - 10 | 1059 | 86 | ||
| AK087935.1 | - | 9 - 10 | 86 | |||
| BC051540.1 | - | 8 - 9 | 86 | |||
| BC096051.1 | - | 9 - 10 | 843 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt3
Genomic Map