Assay Details
| Gene Symbol: Fsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183178.2 | NP_899001.1 | 11 - 12 | 1419 | 76 | |
| GenBank mRNA | AK140132.1 | - | 11 - 12 | 1453 | 76 | |
| AK144257.1 | - | 11 - 12 | 1354 | 76 | ||
| BC055106.1 | - | 11 - 12 | 1419 | 6532001 | 76 | |
More Information
Set Membership: |
|

Fsd1
Genomic Map