Assay Details
| Gene Symbol: Fsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183178.2 | NP_899001.1 | 12 - 13 | 1504 | 78 | |
| GenBank mRNA | AK140132.1 | - | 12 - 13 | 1538 | 78 | |
| AK144257.1 | - | 12 - 13 | 1439 | 78 | ||
| BC055106.1 | - | 12 - 13 | 1504 | 6532001 | 78 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377693 | BC055106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377693 | NM_183178.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377693 | AK140132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377693 | AK144257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377693 | BC055106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377693 | NM_183178.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377693 | AK140132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P377693 | AK144257.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637283 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71190 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77223 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112947 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165421 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377693 | BC055106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377693 | NM_183178.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377693 | AK140132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377693 | AK144257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377693 | BC055106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377693 | NM_183178.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377693 | AK140132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P377693 | AK144257.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253127 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165421 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112947 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08810075 | - | - |
| Whole Mouse Genome G4122A | A_52_P377693 | BC055106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377693 | NM_183178.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377693 | AK140132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377693 | AK144257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377693 | BC055106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377693 | NM_183178.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377693 | AK140132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P377693 | AK144257.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114326 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112947 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13326652 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13112906 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13112904 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13326650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13112907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13201882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13112906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13067876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13326651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13152578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13112904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13387523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13441577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13081464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13186242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13201887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13081465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253141 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10446002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446012 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446008 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435421_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10446001 | - | - |
| Mouse Genome 430 2.0 Array | 1435421_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435421_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240121_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240121_at | - | - |
| Mouse Expression Set 430 | 1435421_PM_at | - | - |
| Mouse Expression Set 430 | 1435421_at | - | - |
| Murine Genome U74v2 Set | 137983_PM_at | - | - |
| Murine Genome U74v2 Set | 137983_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435421_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435421_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 661908 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6940576 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940576 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254128 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254128 | - | - |
| mouse-6 v1.1 (Illumina) | scl50605.12.1_93-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940576 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50605.12.1_93-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940576 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1254128 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254128 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50605.12.1_93-S | - | - |

Fsd1
Genomic Map