Assay Details
| Gene Symbol: Shd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159523.1 | NP_001152995.1 | 4 - 5 | 855 | 66 | |
| NM_009168.2 | NP_033194.1 | 4 - 5 | 852 | 66 | ||
| GenBank mRNA | AB018423.1 | - | 4 - 5 | 851 | 66 | |
| AK142491.1 | - | 4 - 5 | 868 | 66 | ||
| AK164639.1 | - | 4 - 5 | 788 | 66 | ||
| BC052026.1 | - | 3 - 4 | 1782 | 66 | ||
| BC079574.1 | - | 3 - 4 | 1782 | 66 | ||
| BC103798.1 | - | 4 - 5 | 748 | 30288676 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shd
Genomic Map