Assay Details
| Gene Symbol: Gm30225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076935.1 | - | 3 - 4 | 272 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | low density lipoprotein-related protein 1B (deleted in tumors) | Lrp1b |
9630004P12Rik, LRP-DIT |
-477.0kb | Anti-sense | Chr. 2 - 40595290 - 42653631 on NCBI Build GRCm38 |
| Downstream | kynureninase (L-kynurenine hydrolase) | Kynu | 4432411A05Rik | 423.6kb | Sense | Chr. 2 - 43555258 - 43683048 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm30225
Genomic Map