Assay Details
| Gene Symbol: Mbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290656.1 | NP_001277585.1 | 3 - 4 | 594 | 76 | |
| NM_029924.2 | NP_084200.1 | 10 - 11 | 1658 | 76 | ||
| GenBank mRNA | AK155574.1 | - | 10 - 11 | 1661 | 76 | |
| AK173176.1 | - | 3 - 4 | 525 | 76 | ||
| BC138375.1 | - | 3 - 4 | 499 | 8861395 | 76 | |
| BC138378.1 | - | 3 - 4 | 499 | 8861398 | 76 | |
| CB522230.1 | - | 3 - 4 | 372 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbd5
Genomic Map