Assay Details
| Gene Symbol: Mbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290656.1 | NP_001277585.1 | 8 - 9 | 4542 | 105 | |
| NM_029924.2 | NP_084200.1 | 15 - 16 | 6296 | 105 | ||
| GenBank mRNA | AK173176.1 | - | 8 - 9 | 5163 | 105 | |
| BC050251.1 | - | 4 - 5 | 3054 | 105 | ||
| BC138375.1 | - | 8 - 9 | 4447 | 8861395 | 105 | |
| BC138378.1 | - | 8 - 9 | 4447 | 8861398 | 105 | |
| BU610422.1 | - | 2 - 3 | 391 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbd5
Genomic Map