Assay Details
| Gene Symbol: Gzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028986.3 | NP_083262.1 | 1 - 2 | 91 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_028986.3 transcripts.
Set Membership: |
|

Gzf1
Genomic Map