Assay Details
| Gene Symbol: Cst9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009979.1 | NP_034109.1 | 2 - 3 | 404 | 68 | |
| GenBank mRNA | AB017157.1 | - | 2 - 3 | 379 | 68 | |
| AK006429.1 | - | 2 - 3 | 396 | 68 | ||
| BC048486.1 | - | 2 - 3 | 393 | 6772736 | 68 | |
| Y18243.1 | - | 2 - 3 | 404 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | AB017157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | NM_009979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | AK006429.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | BC048486.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | Y18243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | AB017157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | NM_009979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | AK006429.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | BC048486.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506297 | Y18243.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122757 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150700 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | AB017157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | NM_009979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | AK006429.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | BC048486.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | Y18243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | AB017157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | NM_009979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | AK006429.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | BC048486.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506297 | Y18243.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | AB017157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | NM_009979.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | AK006429.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | BC048486.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | Y18243.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | AB017157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | NM_009979.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | AK006429.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | BC048486.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P506297 | Y18243.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732446 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150700 | - | - |
| Whole Mouse Genome G4122A | A_51_P506297 | AB017157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | NM_009979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | AK006429.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | BC048486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | Y18243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | AB017157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | NM_009979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | AK006429.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | BC048486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P506297 | Y18243.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139071 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150700 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01965185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01499130 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01575000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01965182 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01965184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01499130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01368176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01450038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01965182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01575000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01965184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01602879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01602881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01602882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02179712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02179717 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10476925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476926 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449285_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10476924 | - | - |
| Mouse Genome 430 2.0 Array | 1449285_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449285_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13013_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13013_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200017432.mm.1 | NM_009979.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017432.mm.1 | NM_009979.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017432.mm.1 | TC0200002203.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449285_PM_at | - | - |
| Mouse Expression Set 430 | 1449285_at | - | - |
| Murine Genome U74v2 Set | 103296_at | - | - |
| Murine Genome U74v2 Set | 103296_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449285_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449285_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449285_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449285_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2230706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002230706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239816 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239816 | - | - |
| mouse-6 v1.1 (Illumina) | scl20152.3.1_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2230706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20152.3.1_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2230706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239816 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239816 | - | - |

Cst9
Genomic Map