Assay Details
| Gene Symbol: Trp53i11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025246.1 | NP_001020417.1 | 5 - 6 | 495 | 70 | |
| GenBank mRNA | AK142316.1 | - | 6 - 7 | 668 | 70 | |
| BC024710.1 | - | 4 - 5 | 464 | 70 | ||
| BC048398.1 | - | 5 - 6 | 407 | 6397829 | 70 | |
| BC059041.1 | - | 5 - 6 | 367 | 70 | ||
| BC098206.1 | - | 5 - 6 | 495 | 6812412 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53i11
Genomic Map