Assay Details
| Gene Symbol: Trp53i11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025246.1 | NP_001020417.1 | 6 - 7 | 589 | 85 | |
| GenBank mRNA | AK142316.1 | - | 7 - 8 | 762 | 85 | |
| BC036165.1 | - | 1 - 2 | 72 | 85 | ||
| BC048398.1 | - | 6 - 7 | 501 | 6397829 | 85 | |
| BC098206.1 | - | 6 - 7 | 589 | 6812412 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp53i11
Genomic Map