Assay Details
| Gene Symbol: Prdm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177536.1 | NP_001171007.1 | 2 - 3 | 288 | 83 | |
| GenBank mRNA | AK078754.1 | - | 1 - 2 | 143 | 83 | |
| CJ069425.1 | - | 1 - 2 | 136 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdm11
Genomic Map